Learn More
TMED9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21620-1-AP-150UL
 View more versions of this product
Product Code. 16801225
  • £349.00 / 150µL
Description
TMED9 (also named as GMP25, GP25L2 or p25) is a member of the EMP24/GP25L family. TMED9 probably forms hetero-oligomeric complexes with TMED7 (p27), TMED2 (p24), and TMED10 (p23). Located in the endoplasmic reticulum and the Golgi, TMED9 appears to be involved in vesicular protein trafficking, mainly in the early secretory pathway.
Specifications
Specifications
TMED9
Polyclonal
Unconjugated
TMED9
Glycoprotein 25L2, GMP25, GP25L2, HSGP25L2G, TMED9
Rabbit
Antigen Affinity Chromatography
RUO
361207, 54732, 67511
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5I0E7, Q99KF1, Q9BVK6
Tmed9
TMED9 Fusion Protein Ag13813
150 μL
Primary
Human, Mouse, Rat