missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMED4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £307.00
Specifications
| Antigen | TMED4 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-3 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18097575
|
Novus Biologicals
NB100-56393SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18057855
|
Novus Biologicals
NB100-56393 |
0.1 mg |
£307.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMED4 Polyclonal specifically detects TMED4 in Human samples. It is validated for Western Blot.Specifications
| TMED4 | |
| Western Blot 1-3 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| Endoplasmic reticulum stress-response protein 25, endoplasmic reticulum stress-response protein 25 kDa, ERS25, HNLF, Putative NF-kappa-B-activating protein 156, putative NFkB activating protein HNLF, transmembrane emp24 domain-containing protein 4, transmembrane emp24 protein transport domain containing 4 | |
| TMED4 | |
| IgG | |
| Protein G purified | |
| An approx. 26 kDa protein is recognized in HL60 cell lysate. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q7Z7H5 | |
| 222068 | |
| A synthetic peptide containing amino acids CNYRTQMWDKQKEV (amino aicds 55-67) of human HNLF. Genbank accession no. EAL23751. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title