missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMED4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56393SS
This item is not returnable.
View return policy
Description
TMED4 Polyclonal specifically detects TMED4 in Human samples. It is validated for Western Blot.
Specifications
| TMED4 | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| Endoplasmic reticulum stress-response protein 25, endoplasmic reticulum stress-response protein 25 kDa, ERS25, HNLF, Putative NF-kappa-B-activating protein 156, putative NFkB activating protein HNLF, transmembrane emp24 domain-containing protein 4, transmembrane emp24 protein transport domain containing 4 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 222068 | |
| Human | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1-3 ug/ml | |
| Q7Z7H5 | |
| TMED4 | |
| A synthetic peptide containing amino acids CNYRTQMWDKQKEV (amino aicds 55-67) of human HNLF. Genbank accession no. EAL23751. | |
| 0.025 mg | |
| Primary | |
| An approx. 26 kDa protein is recognized in HL60 cell lysate. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction