Learn More
TMBIM6 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26782-1-AP-150UL
Description
The TMBIM6 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human TMBIM6. This antibody recognizes human, mouse antigen. The TMBIM6 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
| TMBIM6 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 7009 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
| 467 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |