missing translation for 'onlineSavingsMsg'
Learn More

TMBIM6 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Product Code. p-7224164 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16865145 20 μL 20µL
16855145 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16865145 Supplier Proteintech Supplier No. 267821AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The TMBIM6 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human TMBIM6. This antibody recognizes human, mouse antigen. The TMBIM6 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.

Specifications

Antigen TMBIM6
Applications Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA
Classification Polyclonal
Concentration 467 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 7009
Target Species Human, Mouse
Isotype IgG
Show More Show Less
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.