missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress TMA-DPH
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Content And Storage | -20°C, sealed storage, away from moisture and light ∣ The compound is unstable in solutions, freshly prepared is recommended. |
| Product Type | Dye Reagents |
| Quantity | 1 mg |
| Shipping Condition | Room temperature |
| Target | HBV |
| Molecular Weight (g/mol) | 461.62 g/mol |
| CAS | 115534-33-3 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction