missing translation for 'onlineSavingsMsg'
Learn More

TM4SF19 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

£139.00 - £274.00

Specifications

Antigen TM4SF19
Dilution Western Blot 1:500 - 1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18393797
View Documents
Novus Biologicals
NBP3-16026-20UL
20 μg
£139.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18301337
View Documents
Novus Biologicals
NBP3-16026-100UL
100 μg
£274.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TM4SF19 Polyclonal antibody specifically detects TM4SF19 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

TM4SF19
Western Blot
Unconjugated
Rabbit
Human, Mouse
Osteoclast maturation-associated gene 4 protein, Tetraspan membrane protein OCTM4, transmembrane 4 L six family member 19, transmembrane 4 L6 family member 19
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human TM4SF19 (NP_001191827.1). LLGRHAMLGTGLWGGGLMVLTALLSGGLALLGALICFVTSGVALKDGPFCMFDVSSFNQTQAWKYGYPFKDLHSRNYLYDRSLWNSVCLEPSAAVVWHVSL
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500 - 1:2000
Polyclonal
Purified
RUO
PBS, 50% glycerol, pH7.3
116211
IgG
Affinity purified
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.