missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
£167.00 - £302.00
Specifications
| Antigen | TLR7 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry, Immunohistochemistry reported in scientific literature (PMID 28028829), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24886842), Immunohistochemistry-Paraffin 1:100 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18004575
|
Novus Biologicals
NB100-56682SS |
0.025 mL |
£167.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18787693
|
Novus Biologicals
NB100-56682 |
0.1 mL |
£302.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLR7 Polyclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TLR7 | |
| Western Blot 1:500-1:1000, Flow Cytometry, Immunohistochemistry reported in scientific literature (PMID 28028829), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24886842), Immunohistochemistry-Paraffin 1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| NP_057646 | |
| 51284 | |
| This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 465-483 of human TLR7. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/ml | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity | |
| PBS with 0.05% Sodium Azide | |
| toll-like receptor 7 | |
| TLR7 | |
| IgG | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title