missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB100-56682SS
This item is not returnable.
View return policy
Description
TLR7 Polyclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TLR7 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| toll-like receptor 7 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/ml | |
| Western Blot 1:500-1:1000, Flow Cytometry, Immunohistochemistry reported in scientific literature (PMID 28028829), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24886842), Immunohistochemistry-Paraffin 1:100 | |
| NP_057646 | |
| TLR7 | |
| This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 465-483 of human TLR7. | |
| 0.025 mL | |
| Immunology, Innate Immunity | |
| 51284 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction