missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR4 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TLR4 |
|---|---|
| Concentration | 550 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
Description
The TLR4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse TLR4. This antibody recognizes mouse, human antigen. The TLR4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TLR4 | |
| Western Blot, ELISA | |
| Rabbit | |
| 21898 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 550 μg/mL | |
| Liquid | |
| Mouse, Human | |
| Lps, Ly87, Ran/M1, Rasl2 8, Tlr4, toll like receptor 4 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title