Learn More
TLR4 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30400-1-AP-150UL
This item is not returnable.
View return policy
Description
The TLR4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse TLR4. This antibody recognizes mouse, human antigen. The TLR4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TLR4 | |
| 550 μg/mL | |
| Rabbit | |
| Primary | |
| Mouse, Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| Lps, Ly87, Ran/M1, Rasl2 8, Tlr4, toll like receptor 4 | |
| 150 μL | |
| 21898 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |