missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR4 Antibody (76B357.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56566
This item is not returnable.
View return policy
Description
TLR4 Monoclonal specifically detects TLR4 in Human, Mouse, Rat, Porcine, Bovine, Mammal samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Dot Blot, SDS-Page, Flow (Cell Surface), Flow (Intracellular), ELISA Capture (Matched Antibody Pair), CyTOF-ready, Knockout Validated, Knockdown Validated, Block/Neutralize, Dual RNAscope ISH-IHC.
Specifications
| TLR4 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1-3 ug/ml, Flow Cytometry, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Immunohistochemistry-Frozen 1:500, In vitro assay, Dot Blot, SDS-Page reported in scientific literature (PMID 33166339), Flow (Cell Surface), Flow (Intracellular), ELISA Capture (Matched Antibody Pair) 0.5 ug, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready, Knockout Validated, KnockDown Validated, Block/Neutralize, Dual RNAscope ISH-IHC | |
| 76B357.1, ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, tlr4 76B357.1, TLR4 facs, TLR4 flow cytometry, TLR4 human, TLR4 ihc, TLR4 western blot, TOLL, toll-like receptor 4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used has 100% identity in human, chimp, baboon, Chinese hamster, and rat; 93% identity in bovine and pig; 85% identity in sheep and 78% identity in cat, horse and mouse. Human, Mouse, Rat. Ground squirrel reactivity reported by a customer review. Predicted cross-reactivity based on sequence identity: Chimp (100%), Baboon (100%), Chinese Hamster (100%), Bovine (93%), Pig (93%). Mammal reactivity reported in scientific literature (PMID: 25130694). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoassay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), In vitro Assay, KnockDown, Neutralization | |
| 76B357.1 | |
| Unconjugated | |
| O00206 | |
| TLR4 | |
| This antibody was developed against a portion of amino acids 100-200 of human TLR4 (NP_612564). | |
| 0.1 mg | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| 7099 | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction