missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£175.00 - £308.00
Specifications
| Antigen | TLR2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 28819109) |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18088375
|
Novus Biologicals
NB100-56573SS |
0.025 mg |
£175.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18768113
|
Novus Biologicals
NB100-56573 |
£308.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
TLR2 Polyclonal specifically detects TLR2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TLR2 | |
| Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 28819109) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| NP_003255 | |
| 7097 | |
| This antibody was developed against an extracellular domain of mouse TLR2 (amino acids 564-580 and 751-770). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity, Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| TLR2 | |
| IgG | |
| Protein G purified | |
| An 86 kDa band is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title