Learn More
TLR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NB100-56573
 View more versions of this product
Product Code. 18768113
  • £308.00 / 0.10mg
Description
TLR2 Polyclonal specifically detects TLR2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2
RUO
Purified
Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 28819109)
Rabbit
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.