missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00
Specifications
| Antigen | TLR11 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
TLR11 Polyclonal specifically detects TLR11 in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| TLR11 | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Mouse | |
| NP_991388 | |
| 442887 | |
| Antibody was raised against a peptide corresponding to 15 amino acids near the carboxy terminus of mouse TLR11. The immunogen is located within the last 50 amino acids of TLR11. Amino Acid Squence: CEQLKRRLSKAGQER | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| hgnc31753, Toll like receptor 11 | |
| TLR11 | |
| IgG | |
| Ion exchange chromatography |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title