missing translation for 'onlineSavingsMsg'
Learn More

TLR11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-77204-0.025mg

 View more versions of this product

Product Code. 18712934

  • £134.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TLR11 Polyclonal specifically detects TLR11 in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TLR11
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
hgnc31753, Toll like receptor 11
Rabbit
Ion exchange chromatography
RUO
442887
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
1 mg/mL
Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL
NP_991388
TLR11
Antibody was raised against a peptide corresponding to 15 amino acids near the carboxy terminus of mouse TLR11. The immunogen is located within the last 50 amino acids of TLR11. Amino Acid Squence: CEQLKRRLSKAGQER
0.025 mg
Primary
Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only