Learn More
TLE1 Mouse, anti-Human, CoraLite Plus 750, Clone: 2F2H11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL750-66216-20UL
Description
The TLE1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TLE1. This antibody recognizes human antigen. The TLE1 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 750.
Specifications
| TLE1 | |
| 2F2H11 | |
| CoraLite Plus 750 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG2b |
| Western Blot | |
| 1000 μg/mL | |
| E(Sp1) homolog, ESG, ESG1, GRG1, TLE1 | |
| 20 μL | |
| 7088 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |