missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE1 Mouse, anti-Human, CoraLite Plus 750, Clone: 2F2H11, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | TLE1 |
|---|---|
| Clone | 2F2H11 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Conjugate | CoraLite Plus 750 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18070466
|
Proteintech
CL750-66216-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18050306
|
Proteintech
CL750-66216-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TLE1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TLE1. This antibody recognizes human antigen. The TLE1 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 750.
Specifications
| TLE1 | |
| 1000 μg/mL | |
| CoraLite Plus 750 | |
| Mouse | |
| 7088 | |
| IgG2b | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 2F2H11 | |
| Western Blot | |
| Liquid | |
| Human | |
| E(Sp1) homolog, ESG, ESG1, GRG1, TLE1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title