missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TJP3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifications
| Antigen | TJP3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TJP3 Polyclonal antibody specifically detects TJP3 in Human samples. It is validated for Western BlotSpecifications
| TJP3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| MGC119546, Tight junction protein 3, tight junction protein 3 (zona occludens 3), ZO-3, ZO3tight junction protein ZO-3, zona occludens 3, Zona occludens protein 3, Zonula occludens protein 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 710-919 of human TJP3 (NP_001254489.1). RQWLAPASRRSTRRLYAQAQKLRKHSSHLFTATIPLNGTSDTWYQELKAIIREQQTRPIWTAEDQLDGSLEDNLDLPHHGLADSSADLSCDSRVNSDYETDGEGGAYTDGEGYTDGEGGPYTDVDDEPPAPALARSSEPVQADESQSPRDRGRISAHQGAQVDSRHPQGQWRQDSMRTYEREALKKKFMRVHDAESSDEDGYDWGPATDL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 27134 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title