missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TJP3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94303-0.1ml
This item is not returnable.
View return policy
Description
TJP3 Polyclonal antibody specifically detects TJP3 in Human samples. It is validated for Western Blot
Specifications
| TJP3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MGC119546, Tight junction protein 3, tight junction protein 3 (zona occludens 3), ZO-3, ZO3tight junction protein ZO-3, zona occludens 3, Zona occludens protein 3, Zonula occludens protein 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 710-919 of human TJP3 (NP_001254489.1). RQWLAPASRRSTRRLYAQAQKLRKHSSHLFTATIPLNGTSDTWYQELKAIIREQQTRPIWTAEDQLDGSLEDNLDLPHHGLADSSADLSCDSRVNSDYETDGEGGAYTDGEGYTDGEGGPYTDVDDEPPAPALARSSEPVQADESQSPRDRGRISAHQGAQVDSRHPQGQWRQDSMRTYEREALKKKFMRVHDAESSDEDGYDWGPATDL | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 27134 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction