missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF25 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80034
This item is not returnable.
View return policy
Description
KIF25 Polyclonal specifically detects KIF25 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| KIF25 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| kinesin family member 25 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3834 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| NP_085118 | |
| KIF25 | |
| Synthetic peptide directed towards the C terminal of human KIF25. Peptide sequence VLGALLEHRGHAPYRNSRLTHLLQDCLGGDAKLLVILCISPSQRHLAQTL. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Canine 79%, Rat 79%. | |
| Human, Rat, Canine, Equine, Guinea Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction