missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) stearate, Thermo Scientific™

Product Code. 11324537
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324537 100 g 100g
11334537 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324537 Supplier Thermo Scientific Alfa Aesar Supplier No. 039605.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6994-59-8
Molecular Formula C36H70O4Sn
Molecular Weight (g/mol) 685.662
MDL Number MFCD00050720
InChI Key CYCFYXLDTSNTGP-UHFFFAOYSA-L
Synonym tin distearate, pure, tin ii stearate, stannous stearate, octadecanoate; tin 2+, acmc-1bh23, distearic acid tin ii salt, tin, ion sn2+ bis formula 300
PubChem CID 2734723
IUPAC Name octadecanoate;tin(2+)
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Sn+2]

Specifications

Quantity 100 g
UN Number UN3146
Solubility Information Insoluble in water.
Formula Weight 685.69
Physical Form Chunks and/or Powder
Chemical Name or Material Tin(II) stearate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.