accessibility menu, dialog, popup

UserName

SnH3COOH3COO

CAS RN 6994-59-8

IUPAC Name: λ²-tin(2+) dioctadecanoate
Synonyms: λ²-tin(2+) bis(n-octadecanoate)
Molecular Weight (g/mol): 685.66
Molecular Formula: C36H70O4Sn
InChi Key: CYCFYXLDTSNTGP-UHFFFAOYSA-L
SMILES: [Sn++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results