missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TIMP-1 Antibody (2A5), Novus Biologicals™
Mouse Monoclonal Antibody
£162.00 - £402.00
Specifications
| Antigen | TIMP-1 |
|---|---|
| Clone | 2A5 |
| Dilution | Western Blot : 1-2 μg/mL, ELISA, Immunohistochemistry-Paraffin : 1-5 μg/mL |
| Applications | ELISA, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066826
|
Novus Biologicals™
NBP3-30888-200ug |
200 μg |
£402.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067077
|
Novus Biologicals™
NBP3-30888-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIMP-1 Monoclonal antibody specifically detects TIMP-1 in Human,Macaque samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)Specifications
| TIMP-1 | |
| Western Blot : 1-2 μg/mL, ELISA, Immunohistochemistry-Paraffin : 1-5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Macaque | |
| CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1 | |
| Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the human TIMP-1 protein and mouse myeloma Ag8563 cells. Sequence common in monkey | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 2A5 | |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix | |
| Phosphate buffered saline | |
| 7076 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title