missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TIMP-1 Antibody (2A5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30888-50ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TIMP-1 Monoclonal antibody specifically detects TIMP-1 in Human,Macaque samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Spezifikation
| TIMP-1 | |
| Monoclonal | |
| Unconjugated | |
| Phosphate buffered saline | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Macaque | |
| Purified |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| 2A5 | |
| Western Blot : 1-2 μg/mL, ELISA, Immunohistochemistry-Paraffin : 1-5 μg/mL | |
| CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1 | |
| Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the human TIMP-1 protein and mouse myeloma Ag8563 cells. Sequence common in monkey | |
| 50 μg | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix | |
| 7076 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur