missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-3, Mouse anti-Human, Clone: TIM3/3113, Novus Biologicals™
Mouse Monoclonal Antibody
£224.00 - £395.00
Specifications
| Antigen | TIM-3 |
|---|---|
| Clone | TIM3/3113 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18253978
|
Novus Biologicals
NBP2-79702-20UG |
20 μg |
£224.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18233446
|
Novus Biologicals
NBP2-79702-100UG |
100 μg |
£395.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| TIM-3 | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Immunology, Innate Immunity | |
| 10 mM PBS with 0.05% BSA | |
| CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
| HAVCR2 | |
| IgG2a κ | |
| Protein A or G purified |
| TIM3/3113 | |
| ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q8TDQ0, Q8TDQ0 | |
| 84868 | |
| Recombinant fragment of human TIM-3 protein (around aa 22-202) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title