missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-3, Mouse anti-Human, Clone: TIM3/3113, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79702-20UG
This item is not returnable.
View return policy
Description
TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| TIM-3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Q8TDQ0, Q8TDQ0 | |
| HAVCR2 | |
| Recombinant fragment of human TIM-3 protein (around aa 22-202) (exact sequence is proprietary) | |
| 20 μg | |
| Immunology, Innate Immunity | |
| 84868 | |
| Store at 4°C. | |
| IgG2a κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| TIM3/3113 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction