missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-3 (CD366) Mouse anti-Human, BB515, Clone: 7D3, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 565569
This item is not returnable.
View return policy
Description
TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.
TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.
Specifications
| TIM-3 (CD366) | |
| Monoclonal | |
| Brilliant Blue 515 (BB515) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 25 Tests | |
| Cell Biology | |
| Human | |
| Purified |
| Flow Cytometry | |
| 7D3 | |
| Vol. Per test: 5μL | |
| CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3 | |
| Human TIM-3 | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction