missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-3 (CD366) Mouse anti-Human, BB515, Clone: 7D3, BD Horizon™
Mouse Monoclonal Antibody
£206.78 - £388.72
Specifications
| Antigen | TIM-3 (CD366) |
|---|---|
| Clone | 7D3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 515 (BB515) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15894399
|
BD Biosciences
565569 |
25 Tests |
£217.00 £206.78 / Each Save £10.22 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15884399
|
BD Biosciences
565568 |
100 Tests |
£393.00 £388.72 / Each Save £4.28 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.
TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.Specifications
| TIM-3 (CD366) | |
| Flow Cytometry | |
| Brilliant Blue 515 (BB515) | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human TIM-3 | |
| Primary | |
| Vol. Per test: 5μL |
| 7D3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title