missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGIT, Mouse anti-Human, Clone: TIGIT/3017, Novus Biologicals™
Mouse Monoclonal Antibody
£226.00 - £410.00
Specifications
| Antigen | TIGIT |
|---|---|
| Clone | TIGIT/3017 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1-2 ug/million cells, ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18034834
|
Novus Biologicals
NBP2-79793-20UG |
20 μg |
£226.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18285677
|
Novus Biologicals
NBP2-79793-100UG |
100 μg |
£410.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIGIT Monoclonal specifically detects TIGIT in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| TIGIT | |
| 0.2 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM | |
| TIGIT | |
| IgG2c κ | |
| Protein A or G purified |
| TIGIT/3017 | |
| Flow Cytometry 1-2 ug/million cells, ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 201633 | |
| Recombinant fragment (around aa 22-141) of human TIGIT protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title