missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGIT, Mouse anti-Human, Clone: TIGIT/3017, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79793-20UG
This item is not returnable.
View return policy
Description
TIGIT Monoclonal specifically detects TIGIT in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| TIGIT | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1-2 ug/million cells, ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 201633 | |
| Store at 4°C. | |
| IgG2c κ |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| TIGIT/3017 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| TIGIT | |
| Recombinant fragment (around aa 22-141) of human TIGIT protein (exact sequence is proprietary) | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction