missing translation for 'onlineSavingsMsg'
Learn More

Thyroid Peroxidase, Mouse anti-Human, Clone: TPO/1922, Novus Biologicals™

Mouse Monoclonal Antibody

£203.00 - £377.00

Specifications

Antigen Thyroid Peroxidase
Clone TPO/1922
Concentration 0.2 mg/mL
Dilution Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18287866
View Documents
Novus Biologicals
NBP2-79789-20UG
20 μg
£203.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18225767
View Documents
Novus Biologicals
NBP2-79789-100UG
100 μg
£377.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thyroid Peroxidase Monoclonal specifically detects Thyroid Peroxidase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

Thyroid Peroxidase
0.2 mg/mL
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Unconjugated
Mouse
Lipid and Metabolism
10 mM PBS with 0.05% BSA
EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8
TPO
IgG1 κ
Protein A or G purified
Malignant thyroid tumors exhibit an anomaly in Thyroid Peroxidase resulting in lower affinity for anti-Thyroid Peroxidase. Therefore, this antibody may aid in the differentiation between benign and malignant thyroid tumors.
TPO/1922
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Monoclonal
Purified
RUO
Human
P07202
7173
Recombinant fragment of human Thyroid Peroxidase(around aa 685-804) (Exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.