missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroid Peroxidase, Mouse anti-Human, Clone: TPO/1922, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79789-20UG
This item is not returnable.
View return policy
Description
Thyroid Peroxidase Monoclonal specifically detects Thyroid Peroxidase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Thyroid Peroxidase | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P07202 | |
| TPO | |
| Recombinant fragment of human Thyroid Peroxidase(around aa 685-804) (Exact sequence is proprietary) | |
| 20 μg | |
| Lipid and Metabolism | |
| 7173 | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| TPO/1922 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Malignant thyroid tumors exhibit an anomaly in Thyroid Peroxidase resulting in lower affinity for anti-Thyroid Peroxidase. Therefore, this antibody may aid in the differentiation between benign and malignant thyroid tumors. | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction