missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymidine Kinase 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79890
This item is not returnable.
View return policy
Description
Thymidine Kinase 2 Polyclonal specifically detects Thymidine Kinase 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Thymidine Kinase 2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 2.7.1.21, MTDPS2, Mt-TK, MTTK, thymidine kinase 2, mitochondrial | |
| Rabbit | |
| 34 kDa | |
| 100 μL | |
| Proteases & Other Enzymes | |
| 7084 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| NP_004605 | |
| TK2 | |
| The specific Immunogen is proprietary information. Peptide sequence HLHEEWLIKGSLFPMAAPVLVIEADHHMERMLELFEQNRDRILTPENRKH. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Pig: 86%; Rat: 86%; Horse: 86%; Mouse: 86%; Guinea pig: 86%; Bovine: 85%; Rabbit: 79%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu