Learn More
DWK Life Sciences Three Part Retaining Device, complete, for stopcock key, threaded, hollow
Shop All DWK Life Sciences ProductsDescription
The 3-part tap safety device holds and secures the hollow DURAN™ tap plug in the tap sleeve.
- 3-part, for hollow tap plugs
- Three part retaining device holds and secures the tap plug in the tap sleeve
Specifications
Specifications
| Material | Borosilicate Glass (3.3), Polypropylene |
| Autoclavable | Autoclavable |
| Closure Size | 14.5 NS |
| No. per Pack | 100/Pk. |
| For Use With (Application) | For securing the hollow DURAN™ tap plug in the ground DURAN™ one-way tap |