missing translation for 'onlineSavingsMsg'
Learn More

THOC3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-57297

 View more versions of this product

Product Code. 18282355

  • £381.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

THOC3 Polyclonal specifically detects THOC3 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

THOC3
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
hTREX45, MGC5469, TEX1 homolog, TEX1THO complex subunit 3, THO complex 3, tho3
Rabbit
Affinity purified
RUO
Primary
This product is specific to Subunit or Isoform: 3.
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
IgG
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
Q96J01
THOC3
Synthetic peptides corresponding to THOC3(THO complex 3) The peptide sequence was selected from the middle region of THOC3. Peptide sequence LWEVQCESPTFTVAWHPKRPLLAFACDDKDGKYDSSREAGTVKLFGLPND.
100 μL
Core ESC Like Genes, Stem Cell Markers
84321
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.