missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Thiophosphate ester Antibody (SR4347), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-27854-100ul

 View more versions of this product

Product Code. 30041713

  • £290.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Thiophosphate ester Monoclonal antibody specifically detects Thiophosphate ester in Nonspecies specific samples. It is validated for Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Thiophosphate ester
Monoclonal
Unconjugated
PBS (pH 7.4), 150mM NaCl, 50% glycerol
A synthesized peptide derived from human Thiophosphate ester
100 μL
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG
Immunoprecipitation
SR4347
Immunoprecipitation 1:1000
Rabbit
Affinity purified
RUO
Non-species specific
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.