missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Thiophosphate ester Antibody (SR4347), Novus Biologicals™
Rabbit Monoclonal Antibody
£240.00 - £290.00
Specifications
| Antigen | Thiophosphate ester |
|---|---|
| Clone | SR4347 |
| Dilution | Immunoprecipitation 1:1000 |
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041713
|
Novus Biologicals™
NBP3-27854-100ul |
100 μL |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041514
|
Novus Biologicals™
NBP3-27854-50ul |
50 μL |
£240.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thiophosphate ester Monoclonal antibody specifically detects Thiophosphate ester in Nonspecies specific samples. It is validated for ImmunoprecipitationSpecifications
| Thiophosphate ester | |
| Immunoprecipitation 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| IgG | |
| Affinity purified |
| SR4347 | |
| Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Non-species specific | |
| A synthesized peptide derived from human Thiophosphate ester | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title