missing translation for 'onlineSavingsMsg'
Learn More

Thiophenol, 99%

Product Code. 10227113 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
1 L
5 L
Packaging:
Glass Bottle
Metal drum
Unit Size:
1 L
25mL
5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10227113 5 L Metal drum 5 L
10416960 25 mL Metal drum 25mL
10593681 500 mL Metal drum 500mL
10715631 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10227113 Supplier Thermo Scientific Acros Supplier No. 138870050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 108-98-5 | C6H6S | 110.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-98-5
MDL Number MFCD00004826
InChI Key RMVRSNDYEFQCLF-UHFFFAOYSA-N
Synonym thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto
PubChem CID 7969
ChEBI CHEBI:48498
IUPAC Name benzenethiol
SMILES C1=CC=C(C=C1)S

Specifications

Melting Point -15.0°C
Color Colorless to Yellow
Density 1.0780g/mL
Boiling Point 167.0°C to 169.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Metal drum
Molecular Formula C6H6S
Linear Formula C6H5SH
Refractive Index 1.5880 to 1.5900
Quantity 5 L
Beilstein 06, 294
Fieser 06,585; 07,367; 09,465; 10,399; 12,485; 13,53; 16,327
Merck Index 15, 9508
Specific Gravity 1.078
Solubility Information Solubility in water: insoluble. Other solubilities: very soluble in alcohol, miscible with ether, benzene and cs2
Molecular Weight (g/mol) 110.17
Viscosity 0.981 mm2/s (30°C)
Formula Weight 110.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Thiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.