missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-3-carboxaldehyde diethyl acetal, 98%, Thermo Scientific™

Product Code. 11388306
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11388306

Brand: Thermo Scientific Alfa Aesar L15520.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Thiophene-3-carboxaldehyde diethyl aceta is used in OLED materials. Also used as electronic intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3199-44-8
Molecular Formula C9H14O2S
Molecular Weight (g/mol) 186.269
MDL Number MFCD01318173
InChI Key VHXDNUQYRTTWTH-UHFFFAOYSA-N
Synonym 3-diethoxymethyl thiophene, thiophene-3-carboxaldehyde diethyl acetal, acmc-1cotb, diethoxy-3-thienylmethane, thiophene,3-diethoxymethyl, 3-thiophenecarboxaldehyde diethyl acetal
PubChem CID 7006660
IUPAC Name 3-(diethoxymethyl)thiophene
SMILES CCOC(C1=CSC=C1)OCC

Specifications

Boiling Point 130°C (30 mmHg)
Refractive Index 1.4865
Quantity 5 g
Beilstein 125429
Solubility Information Insoluble in water.
Formula Weight 186.27
Percent Purity 98%
Chemical Name or Material Thiophene-3-carboxaldehyde diethyl acetal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.