accessibility menu, dialog, popup

UserName

CAS RN 3199-44-8

H3COSOCH3

CAS RN 3199-44-8

IUPAC Name: 3-(diethoxymethyl)thiophene
Synonyms: 3-(diethoxymethyl)thiophene
Molecular Weight (g/mol): 186.27
Molecular Formula: C9H14O2S
InChi Key: VHXDNUQYRTTWTH-UHFFFAOYSA-N
SMILES: CCOC(OCC)C1=CSC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results