missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-3-acetic acid hydrazide, 97%

Product Code. 11348605
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348605 5 g 5g
11338605 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348605 Supplier Thermo Scientific Alfa Aesar Supplier No. L12077.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 175276-94-5 | C6H8N2OS | 156.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175276-94-5
Molecular Formula C6H8N2OS
Molecular Weight (g/mol) 156.20
MDL Number MFCD00052946
InChI Key YNMRDHMMFCGQBD-UHFFFAOYSA-N
Synonym 2-thiophen-3-yl acetohydrazide, thiophene-3-acetic acid hydrazide, 2-3-thienyl acetohydrazide, 2-thien-3-yl acetohydrazide, 3-thiopheneacetic acid,hydrazide, 3 thienyl-acetic acid hydrazide, 2-thien-3-ylacetohydrazide, acmc-20ajii, maybridge1_008350, 3-thienyl acetohydrazide
PubChem CID 468649
IUPAC Name 2-thiophen-3-ylacetohydrazide
SMILES NNC(=O)CC1=CSC=C1

Specifications

Melting Point 85°C to 86°C
Quantity 5 g
Beilstein 120473
Formula Weight 156.21
Percent Purity 97%
Chemical Name or Material Thiophene-3-acetic acid hydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.