CAS RN 175276-94-5
IUPAC Name:
2-(thiophen-3-yl)acetohydrazide
Synonyms:
2-(thiophen-3-yl)acetohydrazide
Molecular Weight (g/mol):
156.2
Molecular Formula:
C6H8N2OS
InChi Key:
YNMRDHMMFCGQBD-UHFFFAOYSA-N
SMILES:
NNC(=O)CC1=CSC=C1
1
–
2
of
2
results
2
Thiophene-3-acetic acid hydrazide, 97%
CAS: 175276-94-5 Molecular Formula: C6H8N2OS Molecular Weight (g/mol): 156.20 MDL Number: MFCD00052946 InChI Key: YNMRDHMMFCGQBD-UHFFFAOYSA-N Synonym: 2-thiophen-3-yl acetohydrazide,thiophene-3-acetic acid hydrazide,2-3-thienyl acetohydrazide,2-thien-3-yl acetohydrazide,3-thiopheneacetic acid,hydrazide,3 thienyl-acetic acid hydrazide,2-thien-3-ylacetohydrazide,acmc-20ajii,maybridge1_008350,3-thienyl acetohydrazide PubChem CID: 468649 IUPAC Name: 2-thiophen-3-ylacetohydrazide SMILES: NNC(=O)CC1=CSC=C1