missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-thiocarboxamide, 97%, Thermo Scientific™

Product Code. 11470738
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11470738

Brand: Thermo Scientific Alfa Aesar B25225.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Thiophene-2-thiocarboxamide is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20300-02-1
Molecular Formula C5H5NS2
Molecular Weight (g/mol) 143.222
MDL Number MFCD00052890
InChI Key HDXYHAPUCGQOBX-UHFFFAOYSA-N
Synonym thiophene-2-thiocarboxamide, 2-thiophenecarbothioamide, thiophene-2-thioamide, thiophene-2-carbothioic acid amide, thiophene,2-thiocarboxamide, amino-2-thienylmethane-1-thione, 2-thienylthioamide, 2-carbamothioylthiophene, acmc-1cfey, 2-thiophenethiocarboxamide
PubChem CID 737237
IUPAC Name thiophene-2-carbothioamide
SMILES C1=CSC(=C1)C(=S)N

Specifications

Melting Point 106°C to 109°C
Quantity 25 g
UN Number UN2811
Beilstein 110159
Solubility Information Insoluble in water.
Formula Weight 143.23
Percent Purity 97%
Chemical Name or Material Thiophene-2-thiocarboxamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.