CAS RN 20300-02-1
CAS RN 20300-02-1
IUPAC Name:
thiophene-2-carbothioamide
Synonyms:
thiophene,2-thiocarboxamide
Molecular Weight (g/mol):
143.22
Molecular Formula:
C5H5NS2
InChi Key:
HDXYHAPUCGQOBX-UHFFFAOYSA-N
SMILES:
NC(=S)C1=CC=CS1
1
–
1
of
1
results
1
Thiophene-2-thiocarboxamide, 97%, Thermo Scientific™
CAS: 20300-02-1 Molecular Formula: C5H5NS2 Molecular Weight (g/mol): 143.222 MDL Number: MFCD00052890 InChI Key: HDXYHAPUCGQOBX-UHFFFAOYSA-N Synonym: thiophene-2-thiocarboxamide,2-thiophenecarbothioamide,thiophene-2-thioamide,thiophene-2-carbothioic acid amide,thiophene,2-thiocarboxamide,amino-2-thienylmethane-1-thione,2-thienylthioamide,2-carbamothioylthiophene,acmc-1cfey,2-thiophenethiocarboxamide PubChem CID: 737237 IUPAC Name: thiophene-2-carbothioamide SMILES: C1=CSC(=C1)C(=S)N