missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiophene-2-thiocarboxamide, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Thiophene-2-thiocarboxamide is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Thiophene-2-thiocarboxamide is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 106°C to 109°C |
| Quantity | 5 g |
| UN Number | UN2811 |
| Beilstein | 110159 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 143.23 |
| Percent Purity | 97% |
| Chemical Name or Material | Thiophene-2-thiocarboxamide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction