missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Thiazolyl Blue

Product Code. 16451579
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451579 1 g 1g
16439457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16451579 Supplier MedChemExpress Supplier No. HY15924
 Restricted Item
Estimated Shipment: 04-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Thiazolyl Blue (MTT) is a colorimetric agent widely used to measure cell proliferation. Thiazolyl Blue (MTT) is reduced from yellow color to purple formazan in living cells.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 1 g
Shipping Condition Room temperature
Solubility DMSO : 25 mg/mL (60.34 mM; ultrasonic and warming and heat to 60°C) ∣ H2O : 0.88 mg/mL (2.12 mM; Need ultrasonic and warming)
Molecular Weight (g/mol) 414.32 g/mol
CAS 298-93-1
Synonym MTT ∣ Thiazolyl Blue Tetrazolium bromide ∣ Methylthiazolyldiphenyl-tetrazolium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.