missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Thiazole Orange

Product Code. 16409895
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16409895 250 mg 250mg
16482409 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16409895 Supplier MedChemExpress Supplier No. HYD0150

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Thiazole orange is an unsymmetrical cyanine dye which can be conjugated to oligonucleotides (ONs) to create fluorogenic hybridisation probes. Thiazole orange can be used for reticulocyte analysis.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Product Type Dye Reagents
Quantity 250 mg
Shipping Condition Room temperature
Solubility DMSO : 25 mg/mL (52.45 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble)
Target HBV
Molecular Weight (g/mol) 476.61 g/mol
CAS 107091-89-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.