missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Versette ClipTips™

Product Code. 11694902
Click to view available options
Model:
12.5μL
125/300μL
1250μL
30μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Tip Style:
Automation
Unit Size:
Pack of 768
Pack of 960
This item is not returnable. View return policy

Product Code. 11694902

Brand: Thermo Scientific™ 5608

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Unique interface locks tip onto Versette automated liquid handling system.

Tips have a clasp or clip that locks in place to ensure an effective seal and will only release when mechanically ejected.

  • Unique design – On or Off only – tips lock in place providing a secure fit and preventing loose, uneven or leaky tips
  • Minimizing cross contamination between samples, the tips reach the bottom of tubes
  • Versatile tip range with three sizes for volumes: 1250μL, 300/125μL and 30/12.5μL
  • Rigid rack allows stable, even tip seating

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 2 to 30 μL
Filtered Filtered
Sterility Sterile
Model 12.5μL
For Use With (Application) On or off tips lock in place providing a secure fit and preventing loose, uneven or leaky tips, minimizing cross contamination between samples, provide stable, straight mounting every time
Format Rack
Tip Style Automation
For Use With (Equipment) Thermo Scientific Versette
Product Line Versette ClipTips
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.