missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ Thermo Scientific SPME Liner
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan™ 09214801
Code : 13
Additional Details : Weight : 0.10000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Thermo Scientific instruments
Specifications
GC Inlet Liner, SPME Liner | |
105 mm | |
8 mm |
1/Pk. | |
0.8 mm | |
Thermo Scientific Trace and Focus GCs |